Kaliumchlorid, EMPROVE® EXPERT Ph. Eur., JP, USP, BP BiopharmaceuticalProduction, SAFC®
Lieferant: Merck
Synonyme:
Potassiumchloride
1.37009.9012
1.37009.9029
1.37009.5000
1.37009.1000
1.37009.9012EA
571
EUR
1.37009.9012
1.37009.9029
1.37009.5000
1.37009.1000
Kaliumchlorid, EMPROVE® EXPERT Ph. Eur., JP, USP, BP BiopharmaceuticalProduction, SAFC®
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 Schmelzpunkt: 773 Dichte: 1.98 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...