, anhydrous, EMPROVE® EXPERT BP, Ph. Eur., USP BiopharmaceuticalProduction, SAFC®
Lieferant: Merck
Synonyme:
di-Kaliumphosphat, di-Kaliumorthophosphat, Kaliumphosphat dibasisch, Dikaliumhydrogenphosphat, sek. Kaliumphosphat
1.37010.5000
1.37010.9012
1.37010.9029
1.37010.5000EA
546
EUR
1.37010.5000
1.37010.9012
1.37010.9029
, anhydrous, EMPROVE® EXPERT BP, Ph. Eur., USP BiopharmaceuticalProduction, SAFC®
di-Kaliumhydrogenphosphat
Formel:
K₂HPO₄ Molecular Weight: 174.18 g/mol Schmelzpunkt: 340 Dichte: 2.3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011383 CAS-Nummer: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...