Natriumchlorid, EMPROVE® EXPERT USP, JP, Ph. Eur., BP für die biopharmazeutische Produktion, SAFC®

Lieferant: Merck

Synonyme: Saline, NaCl, Sea salt, Halite

1.37017.5000 1.37017.9012 1.37017.9029 1.37017.1000
1.37017.5000EA 272 EUR
1.37017.5000 1.37017.9012 1.37017.9029 1.37017.1000
Natriumchlorid, EMPROVE® EXPERT USP, JP, Ph. Eur., BP für die biopharmazeutische Produktion, SAFC®
Natriumchlorid

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1461 °C (1013 hPa)
Schmelzpunkt: 801
Dichte: 2.17 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Bestellen

Spezifikation Testergebnisse

Assay (argentometric; calculated on dried substance) 99.5 - 100.5 %
Identity passes test
Appearance of solution passes test
Acidity or alkalinity passes test
Bromide (Br) ≤ 0.005 %
Ferrocyanides passes test
Iodide (I) passes test
Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water) ≤ 0.01
Phosphate (PO₄) ≤ 0.0025 %
Sulfate (SO₄) ≤ 0.0200 %
Heavy metals (as Pb) ≤ 0.0003 %
Al (Aluminium) ≤ 0.00002 %
As (Arsenic) ≤ 0.0001 %
Ba (Barium) passes test
Ca (Calcium) ≤ 0.002 %
Fe (Iron) ≤ 0.0002 %
K (Potassium) ≤ 0.01 %
Magnesium and alkaline-earth metals (as Ca) ≤ 0.0100 %
Residual solvents (ICH Q3C) excluded by production process
Loss on drying (105 °C, 2 h) ≤ 0.5 %
Endotoxins (kinetic turbidimetric test) ≤ 2.5 I.U./g
Microbiological purity (Total aerobic microbial count (TAMC)) ≤ 100.0 CFU/g
Microbiological purity (Total yeast and mould count (TYMC)) ≤ 100.0 CFU/g
DNases (Exo- and endonucleases) (not detectable) passes test
RNases (not detectable) passes test
Proteases (not detectable) passes test
Nicking Activity (not detectable) passes test
Corresponds to Ph Eur,BP,JP,USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR