Glycerin vegetable, anhydrous, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP BiopharmaceuticalProduction, SAFC®
Lieferant: Merck
Synonyme:
1,2,3-Propantriol, Glycerol
1.37028.2500
1.37028.9025
1.37028.9950
1.37028.9010
1.37028.1000
1.37028.2500EA
231
EUR
1.37028.2500
1.37028.9025
1.37028.9950
1.37028.9010
1.37028.1000
Glycerin vegetable, anhydrous, EMPROVE® EXPERT USP, JP, ACS, Ph. Eur., BP BiopharmaceuticalProduction, SAFC®
Glycerin
Formel:
HOCH₂CH(OH)CH₂OH Molecular Weight: 92.09 g/mol Siedepunkt: 290 Schmelzpunkt: 18.6 Dichte: 1.26 g/cm³ (20 °C) Flammpunkt: 177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...