Kaliumdihydrogenphosphat, kristallisiert, EMPROVE® EXPERT Ph. Eur., BP, JPC, NF für die biopharmazeutische Produktion, SAFC®

Lieferant: Merck

1.37039.5000 1.37039.9029
1.37039.5000EA 510 EUR
1.37039.5000 1.37039.9029
Kaliumdihydrogenphosphat, kristallisiert, EMPROVE® EXPERT Ph. Eur., BP, JPC, NF für die biopharmazeutische Produktion, SAFC®
Kaliumdihydrogenphosphat

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Corresponds to Ph Eur, BP, JPC NF

Formel: KH₂PO₄
Siedepunkt: ∼100 °C (1013 hPa)
Schmelzpunkt: 253 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric, calculated on dried substance) 98.0 - 100.5 %
Assay (alkalimetric; dried substance) ≥ 99.0 %
Identity passes test
Appearance of solution passes test
Insoluble substances ≤ 0.2 %
pH-value (2 %; water) 4.2 - 4.6
pH-value (5 %; water) 4.2 - 4.5
Carbonate passes test
Condensed phosphate passes test
Chloride (Cl) ≤ 0.009 %
Fluoride (F) ≤ 0.0010 %
Sulfate (SO₄) ≤ 0.017 %
Heavy metals (as Pb) ≤ 0.0010 %
Al (Aluminium) ≤ 0.0005 %
As (Arsenic) ≤ 0.0002 %
Fe (Iron) ≤ 0.0010 %
Na (Sodium) ≤ 0.1 %
0 passes test
Pb (Lead) ≤ 0.0004 %
Endotoxins ≤ 2.5 I.U./g
Total aerobic microbial count ≤ 100 CFU/g
Total yeast and mould count ≤ 100 CFU/g
DNases (Exo- and endonucleases) (not detectable) passes test
RNases (not detectable) passes test
Proteases (not detectable) passes test
Residual solvents (ICH Q3C) excluded by manufacturing process
Reducing substances passes test
Loss on drying (105 °C) ≤ 0.5 %
Loss on drying (130 °C) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR