Natriumhydroxid 5 mol/l, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Caustic soda
1.37041.2500
1.37041.9025
1.37041.9200
1.37041.9958
1.37041.2500EA
306
EUR
1.37041.2500
1.37041.9025
1.37041.9200
1.37041.9958
Natriumhydroxid 5 mol/l, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®
Sodium hydroxide
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 1390 Schmelzpunkt: 323 Dichte: ≤1.11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 Merck Index: 14,08627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...