D(+)-Glucose, anhydrous, EMPROVE® EXPERT USP, ACS, Ph. Eur., BP, SAFC®

Lieferant: Merck

Synonyme: Dextrose, Traubenzucker

1.37048.5000 1.37048.9012 1.37048.9029
1.37048.5000EA 343 EUR
1.37048.5000 1.37048.9012 1.37048.9029
D(+)-Glucose, anhydrous, EMPROVE® EXPERT USP, ACS, Ph. Eur., BP, SAFC®
D(+)-Glucose
Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Schmelzpunkt: 146
Dichte: 1.54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calc. on anhydrous substance) 97.5 - 102.0 %
Identity (IR) passes test
Identity (spec. rotation calc. on anhydrous substance) passes test
Identity (HPLC) passes test
Identity (water) passes test
Appearance of solution passes test
Color and clarity of solution passes test
Titrable acid ≤ 0.002 meq/g
Conductivity ≤ 20 µS/cm
Spec. rotation (α25/D) 52.5 - 53.0 °
In water insoluble matter ≤ 0.005 %
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.005 %
Heavy metals (as Pb) ≤ 5 ppm
Fe (Iron) ≤ 5 ppm
Related substances [sum of impurities A (maltose) and B (isomaltose)! ≤ 0.4 %
Related substances [Impurity C (maltotriose)! ≤ 0.2 %
Related substances [Impurity D (fructose)! ≤ 0.15 %
Related substances (unspecified impurities) ≤ 0.10 %
Related substances (Sum of all impurities) ≤ 0.5 %
Dextrin passes test
Soluble starch, sulfite ≤ 15 ppm
Starch passes test
Residual solvents (ICH Q3C) excluded by the manufacturing process
Loss on drying (105 °C) ≤ 0.2 %
Water (according to Karl Fischer) ≤ 1.0 %
Sulfated ash (600 °C) ≤ 0.02 %
Microbiological purity (Total aerobic microbial count (TAMC)) ≤ 100 CFU/g
Microbiological purity (Total yeast and mould count (TYMC)) ≤ 100 CFU/g
Bile tolerant Gram negative Bacteria (absent in 1 g) passes test
Escherichia coli (absent in 1 g) passes test
Pseudomonas aeruginosa (absent in 1 g) passes test
Staphylococcus aureus (absent in 1 g) passes test
Candida albicans (absent in 1 g) passes test
Salmonella ssp (absent in 10 g) passes test
Endotoxins ≤ 10 I.U./g
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur,BP,USP,ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR