Imidazol, EMPROVE® EXPERT ACS

Lieferant: Merck
Gefahr

Synonyme: 1,3-Diaza-2,4-cyclopentadien, Glyoxalamin, 1H-Imidazol, Glyoxalin

1.37098.0100 1.37098.1000 1.37098.9012 1.37098.6025
1.37098.0100EA 264 EUR
1.37098.0100 1.37098.1000 1.37098.9012 1.37098.6025
Imidazol, EMPROVE® EXPERT ACS
Imidazol

Imidazole is a critical buffer substance for bioprocesses to enhance the purity of your target protein. Its most widely used application is the preparation of buffer solutions to operate downstream purification columns, packed with resins.


Designed for higher risk application (subject to robust QC specifications).


Standards: Imidazole Emprove® Expert ACS is manufactured in Europe according to cGMP (based on IPEC - PQG GMP Guide for pharmaceutical excipients 2006) guidelines.

Formel: C₃H₄N₂
Molecular Weight: 68.08 g/mol
Siedepunkt: 256 °C (1013 hPa)
Schmelzpunkt: 90 °C
Dichte: 1.233 g/cm³ (20 °C)
Flammpunkt: 145 °C
MDL: MFCD00005183
CAS-Nummer: 288-32-4
UN: 3263
ADR: 8
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR