HPLC columns, LiChroCART®, Purospher® RP-18 HC
Lieferant: Merck
1.50190.7131 1.50195.7131 1.50201.7131 1.50229.7131 1.50233.7131 1.50234.7131 1.50236.7131 1.50302.7131 1.51432.7131 1.51442.7131 1.51443.7131 1.51448.7131
1.50190.7131EA 0 EUR
1.50190.7131 1.50195.7131 1.50201.7131 1.50229.7131 1.50233.7131 1.50234.7131 1.50236.7131 1.50302.7131 1.51432.7131 1.51442.7131 1.51443.7131 1.51448.7131
HPLC columns, LiChroCART®, Purospher® RP-18 HC
Chromatographiesäulen
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.