Acetonitril mit 0,1% (v/v) Ameisensäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.59002.1000 1.59002.2500 1.59002.4000
1.59002.1000EA 119 EUR
1.59002.1000 1.59002.2500 1.59002.4000
Acetonitril mit 0,1% (v/v) Ameisensäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®
Acetonitril mit 0,1% (v/v) Ameisensäure

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to prefectly meet LC-MS quality requirements.

Siedepunkt: 81,6 °C (1013 hPa)
Flammpunkt: 2 °C
Lagertemperatur: Raumtemperatur
UN: 1648
ADR: 3,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR