Wasser mit 0,1% Essigsäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
1.59007.2500
1.59007.2500EA
59.4
EUR
1.59007.2500
Wasser mit 0,1% Essigsäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®
Wasser mit 0,1% (v/v) Essigsäure
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.