Acetonitril mit 0,1% Trifluoressigsäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
1.59014.2500
1.59014.2500EA
205
EUR
1.59014.2500
Acetonitril mit 0,1% Trifluoressigsäure, LiChrosolv®, Hyper-Qualität für die LC-MS, Supelco®
Acetonitril mit 0,1% (v/v) Trifluoressigsäure
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.
Dichte:
0,786 g/cm3 (20 °C) Flammpunkt: 2 °C Lagertemperatur: Raumtemperatur |
UN:
1648 ADR: 3, |
Spezifikation Testergebnisse
Acid content | 0.095 - 0.105 % (v/v) |
Identity | passes test |
Color | ≤ 10 Hazen |
Al (Aluminium) | ≤ 0.05 ppm |
Ca (Calcium) | ≤ 0.05 ppm |
Fe (Iron) | ≤ 0.05 ppm |
K (Potassium) | ≤ 0.025 ppm |
Mg (Magnesium) | ≤ 0.05 ppm |
Na (Sodium) | ≤ 0.25 ppm |
Gradient grade (at 210 nm) | ≤ 1.0 mAU |
Gradient grade (at 254 nm) | ≤ 0.5 mAU |
Suitable for LC-MS; Intensity of background mass peak based on reserpine (APCI/ESI positive) | ≤ 2 ppb |
Filtered by 0.2 µm filter. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...