Salpetersäure 1 mol/l (1 N), Titripur®, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Nitric acid solution

MQ100

1.60307.1003
1.60307.1003EA 73.9 EUR
1.60307.1003
Salpetersäure 1 mol/l (1 N), Titripur®, Supelco®
Salpetersäure

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: HNO₃
Molecular Weight: 63.01 g/mol
Siedepunkt: 87
Schmelzpunkt: -42
Dichte: 1.51
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
UN: 2031
ADR: 8
Merck Index: 13,06610

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR