Schwefelsäure 1 mol/l (2 N), Titripur®, Supelco®

Lieferant: Merck
Achtung


MQ100

1.60313.1003
1.60313.1003EA 37.8 EUR
1.60313.1003
Schwefelsäure 1 mol/l (2 N), Titripur®, Supelco®
Schwefelsäure

Sulphuric acid c(H₂SO₄) = 1 mol/l (2 N) Titripur®


Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: H₂SO₄
Molecular Weight: 98.08 g/mol
Siedepunkt: 330
Schmelzpunkt: 10.38
Dichte: 1.1 g/cm³ (20 °C)
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
UN: 2796
ADR: 8

Bestellen

Spezifikation Testergebnisse

Form Liquid
Concentration 0,995 - 1,005 mol/l
Measurement uncertainty ±0,003 mol/l
Traceability NIST SRM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR