Bor-Standardlösung, 1000 mg/L B in Wasser (aus B(OH₃)) Certipur® Standard für die ICP, Supelco®
Lieferant: Merck
1.70307.0100
1.70307.0100EA 173 EUR
1.70307.0100
Bor-Standardlösung, 1000 mg/L B in Wasser (aus B(OH₃)) Certipur® Standard für die ICP, Supelco®
Standards Kationischer Standards Bor-Standards
H₃BO₃ in water.
Standards: Rückführbar auf SRM von NIST. The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Lieferumfang: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.