5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)

Lieferant: Biotium

10001-2 10001 10001-1
10001-2EA 2160 EUR
10001-2 10001. 10001-1
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Formel: C₈H₄BrClNNa₂O₄P
MDL: MFCD00036757
CAS-Nummer: 102185-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR