BCIP/NBT Kit
Lieferant: Biotium
This kit contains 100 mg each of BCIP sodium salt (10001) and the oxidant NBT (Nitro blue tetrazolium, 10008).
BCIP is the most widely used chromogenic phosphatase substrate, which forms a dark blue precipitate on enzymatic hydrolysis. It is often used with the oxidant NBT (nitro blue tetrazolium chloride), which facilitates the precipitation. BCIP/NBT produces a dark blue/violet precipitate product that can be mounted with permanent or aqueous mounting medium.
Stored desiccated at ≤ 4 °C upon receipt. Protect components from light, especially when in solution.