5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)

Lieferant: Biotium

Synonyme: BCIP Red, Magenta™ phosphate

10004
10004.EA 250 EUR
10004.
5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)
5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)

BCIP Red (5-Bromo-6-chloro-3-indoxyl phosphate, p-toluidine salt ) is a derivative of BCIP. While BCIP yields a blue precipitating product, BCIP Red produces a red (λmax 565 nm) colored precipitate.

Formel: C₈H₆BrClNO₄P·C₇H₉N
MDL: MFCD00210023
CAS-Nummer: 6769-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a017af61b28c0a9fb853c0fb6020d77ab6283c2&vid=4a017af61b28c0a9fb853c0fb6020d77ab6283c2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR