BCIP Red/NBT Kit
Lieferant: Biotium
10005
10005.EA 304 EUR
10005.
BCIP Red/NBT Kit
BCIP Red/NBT
This kit contains 100 mg each of BCIP Red (10004) and the oxidant NBT (10008).
BCIP Red is a derivative of BCIP. Unlike BCIP, which yields a blue precipitating product, BCIP Red produces a red colored precipitate. It is often used with the oxidant NBT, which facilitates the precipitation.