Allylpalladium(II) chloride dimer (≥ 56,0% Pd)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 12012-95-2
Molecular Formula: C6H10Cl2Pd2
Formula Weight: 365.85
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow
Melting Point: 120° dec.
Solubility: Very soluble in dichloromethane and dichloroethane. Slightly soluble in toluene, insoluble in water
Beilstein Reference: 4124623
MDL No.: MFCD00044874
Formel:
C₆H₁₀Cl₂Pd₂ |
MDL:
MFCD00044874 CAS-Nummer: 12012-95-2 EINECS: 234-579-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...