Abacavir System Suitability Mixtures, Reference Standard
Lieferant: USP
1000500
1000500.EA 1290 EUR
1000500.
Abacavir System Suitability Mixtures, Reference Standard
Standards Organische Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
A mixture containing abacavir sulfate and trans-abacavir
Standards: US Pharmacopea Reference Standard