6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)

Lieferant: Biotium

10006
10006.EA 270 EUR
10006.
6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)
6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)

BCIP Pink (6-Chloro-3-indoxyl phosphate, p-toluidine salt) is a derivative of BCIP. While BCIP yields a blue precipitating product, BCIP Pink produces a pink colored (λmax 540 nm) precipitate.

Formel: C₈H₆ClNO₄P·C₇H₁₀N
Molecular Weight: 354,73 g/mol
MDL: MFCD00216690
CAS-Nummer: 159954-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e563f01819d6c815a4fd63aaf163eda42ca1ea6e&vid=e563f01819d6c815a4fd63aaf163eda42ca1ea6e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR