Diphenylborsäure(2-aminoethyl)ester (2-APB), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine
100065-100
100065.100EA
127
EUR
100065.100
Diphenylborsäure(2-aminoethyl)ester (2-APB), Millipore®
Diphenylborsäure(2-aminoethyl)ester (2-APB)
Formel:
C₁₄H₁₆BNO Molecular Weight: 225.1 g/mol |
MDL:
MFCD00014823 CAS-Nummer: 524-95-8 EINECS: 208-366-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...