BCIP Pink/NBT Kit
Lieferant: Biotium
10007
10007.EA
331
EUR
10007.
BCIP Pink/NBT Kit
BCIP Pink/NBT
This kit contains 100 mg each of BCIP Pink (10006) and the precipitation inducer NBT (10008).
BCIP Pink is a derivative of BCIP. Unlike BCIP, which yields a blue precipitating product, BCIP Pink produces a pink colored precipitate. The oxidant NBT facilitates the precipitation.