X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
Lieferant: Biotium
10016
10016.EA
180
EUR
10016.
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
Formel:
C₂₀H₂₆BrClN₂O₇ Schmelzpunkt: 230 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058543 CAS-Nummer: 114162-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...