β-glucuronidase substrates
Lieferant: Biotium
10020
10018
10019
10017
10025
10020.EA
180
EUR
10020.
10018.
10019.
10017.
10025.
β-glucuronidase substrates
Färbemittel
Bildgebende Farbstoffe (in vivo)
β-glucuronidase substrates are easy to detect stains against the background of plant cells.
White solid soluble in water or DMSO.
Bitte beachten Store at −20 °C and protect from light.