IPTG (Isopropyl-β-D-thiogalactopyranosid)
Lieferant: Biotium
Synonyme:
IPTG
10021
10021-1
10021-2
10021.EA
57.4
EUR
10021.
10021-1
10021-2
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel:
C₉H₁₈O₅S Molecular Weight: 238.31 g/mol Siedepunkt: 402 Schmelzpunkt: 109.5…110.5 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063273 CAS-Nummer: 367-93-1 EINECS: 206-703-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...