Chromogenic Phosphatase Substrate Sampler kit
Lieferant: Biotium
Synonyme: BCIP/BCIP red/BCIP pink/NBT
10022
10022.EA 336 EUR
10022.
Chromogenic Phosphatase Substrate Sampler kit
Chromogenic Phosphatase Substrate Sampler kit
The sampler kit contains 10 mg each of BCIP Na (10001), BCIP Pink (10006), BCIP Red (10004) and 250 mg of NBT (10008). NBT forms a co-precipitant and enhances the BCIP reaction.
BCIP (10001) is the most widely used chromogenic phosphatase substrate, which forms a dark blue precipitate on enzymatic hydrolysis. BCIP Pink (10006) and BCIP Red (10004) are derivatives of BCIP. Unlike BCIP that yields a blue precipitating product, BCIP Pink produces a pink colored precipitate while BCIP Red produces a red colored precipitate. BCIP substrates often used with the oxidant NBT (nitro blue tetrazolium chloride) (10008), which facilitates the precipitation.
The sampler kit contains all three BCIP substrates and NBT for your convenience.