Cäsiumiodid 99,9 Metals basis
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cesiumiodide
10022.14
10022.22
10022.14EA
78
EUR
10022.14
10022.22
Cäsiumiodid 99,9 Metals basis
Cäsiumiodid
Purity: 99.9%
CAS No.: 7789-17-5
Molecular Formula: CsI
Formula Weight: 259.81
Physical Form: -20 Mesh
Refractive Index: 1.7876
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 626°
Boiling Point: 1280°
Solubility: Very soluble in water. Soluble in ethanol. Slightly soluble in methanol. Insoluble in acetone
Merck Reference: 14,2014
MDL No.: MFCD00010962
Formel:
CsI Molecular Weight: 259.81 g/mol Siedepunkt: 1280 °C Schmelzpunkt: 626 °C Dichte: 3.21 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010962 CAS-Nummer: 7789-17-5 EINECS: 232-145-2 Merck Index: 14,2014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...