X-β-D-cellobioside

Lieferant: Biotium
Achtung

Synonyme: 5-Bromo-4-chloro-3-indolyl-β-D-cellobioside

10026
10026.EA 226 EUR
10026.
X-β-D-cellobioside
X-β-D-cellobioside

X-β-D-cellobioside is a chromogenic substrate for β-cellobiosidase.


  • Cellobiosidase Substrate


White solid soluble in DMSO.


Enzymatic cleavage of the substrate generates a dark blue-coloured precipitate.


Bitte beachten Store at −20 °C and protect from light.

Formel: C₂₀H₂₅BrClNO₁₁
Molecular Weight: 570,78 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 177966-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22fbe28aee0097ff208e8e7395618aa36043d75a&vid=cce741967e8f96ea54640b842625c6689dcc6b3b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR