Resazurin Natriumsalz
Lieferant: Biotium
|
Achtung
|
Synonyme:
7-Natriooxy-3H-phenoxazin-3-on-10-oxid
10054
10054.EA
179
EUR
10054.
Resazurin Natriumsalz
Resazurin Natriumsalz
Formel:
C₁₂H₆NNaO₄ Molecular Weight: 251.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005036 CAS-Nummer: 62758-13-8 EINECS: 263-718-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...