10-Acetyl-3,7-dihydroxyphenoxazine Peroxidase-Substrat

Lieferant: Biotium

10061
10061.EA 268 EUR
10061.
10-Acetyl-3,7-dihydroxyphenoxazine Peroxidase-Substrat
10-Acetyl-3,7-dihydroxyphenoxazine

0-Acetyl-3,7-dihydroxyphenoxazine (Amplex® Red), is regarded as the best fluorogenic substrate for peroxidase and H2O2 because it is highly sensitive, specific and stable.


  • Detect HRP in immunoassays
  • Detect trace amounts of hydrogen peroxide (H2O2)
  • Detect using fluorometer or microplate reader
  • λEx/λEm (pH 9)= 571/585 nm (end product)
  • ε= 54,000 (end product)
  • White to off-white solid soluble in DMSO or DMF
  • Store desiccated at -20°C and protect from light


10-Acetyl-3,7-dihydroxyphenoxazine (N-Acetyl-3,7-dihydroxyphenoxazine or ADHP), which has been marketed under the name Amplex® Red, is regarded as the best fluorogenic substrate for peroxidase because it is highly specific and stable. The substrate itself is nearly colorless and nonfluorescent until it is oxidized by H2O2 (reacting in a 1:1 stoichiometry) in the presence of horseradish peroxidase (HRP) to become the highly red fluorescent resorufin. H2O2 is produced by many enzymatic reactions, and therefore ADHP can be used to detect the activity of many different enzymes.

Formel: C₁₄H₁₁NO₄
Lagertemperatur: Gefrierschrank
MDL: MFCD00467861
CAS-Nummer: 119171-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be85df14edf394521c7566d6d73cfc1ed3b2d213&vid=be85df14edf394521c7566d6d73cfc1ed3b2d213&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR