Arsen ≥99% (Metall-Basis), Pulver -20 mesh

Lieferant: Thermo Fisher Scientific
Gefahr

10101.18
10101.18EA 176 EUR
10101.18
Arsen ≥99% (Metall-Basis), Pulver -20 mesh
Arsenic

Purity: 99% CAS No.: 7440-38-2 Appearance: Silver to black Odour: Odourless UN No.: 1558 MDL No.: MFCD00085309

Formel: As
Dichte: 5,7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00085309
CAS-Nummer: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR