Bismut ≥99,5% (Metall-Basis), Pulver -325 mesh

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Wismut

10111.36 10111.22
10111.36EA 222 EUR
10111.36 10111.22
Bismut ≥99,5% (Metall-Basis), Pulver -325 mesh
Bismut

Purity: 99.5% CAS No.: 7440-69-9 Physical Form: Apparent density 3.2-4.0g/cm3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00134033

Formel: Bi
Dichte: 9,78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134033
CAS-Nummer: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR