Graphit 75 - 82% ≥75-82%, mikrokristallines Pulver -300 mesh 18-25% Asche
Lieferant: Thermo Fisher Scientific
|
Achtung
|
10130.14
10130.30
10130.A1
10130.14EA
28.8
EUR
10130.14
10130.30
10130.A1
Graphit 75 - 82% ≥75-82%, mikrokristallines Pulver -300 mesh 18-25% Asche
Graphit
CAS No.: 7782-42-5
Physical Form: Powder
Appearance: Black
Odour: Odourless
UN No.: 1325
MDL No.: MFCD00144065
Formel:
C Schmelzpunkt: 3827 °C Dichte: 2,09…2.23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144065 CAS-Nummer: 7782-42-5 EINECS: 231-955-3 UN: 1325 ADR: 4.1,III Merck Index: 12,04560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...