Lactose Monohydrat, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals

Synonyme: 4-(β-D-Galactopyranosyl)-D-glucopyranose Monohydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose 1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-Monohydrat

101394SEA 34.1 EUR
101394S
Lactose Monohydrat, AnalaR NORMAPUR® analytisches Reagens
Lactose Monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
Molecular Weight: 360,32 g/mol
Schmelzpunkt: 223 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 10039-26-6
EINECS: 200-559-2

Bestellen

Spezifikation Testergebnisse

Specific optical rotation (10 %; water) 52.0 - 52.8 °
Acidity Max. 0.005 meq/g
Ignition residue (SO4) Max. 0.1 %
Insolubility in water Max. 50 ppm
Water 4.8 - 5.4 %
As (Arsenic) Max. 1 ppm
Cu (Copper) Max. 0.5 ppm
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 0.5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR