Chrom ≥99% (Metall-Basis), Pulver -100 mesh

Lieferant: Thermo Fisher Scientific
Achtung

10147.A3 10147.09 10147.22 10147.36
10147.A3EA 1155 EUR
10147.A3 10147.09 10147.22 10147.36
Chrom ≥99% (Metall-Basis), Pulver -100 mesh
Chromium

Purity: 99% CAS No.: 7440-47-3 Physical Form: Apparent density 2.0-3.0g/cm3 Appearance: Silver Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944

Formel: Cr
Lagertemperatur: Raumtemperatur
MDL: MFCD00010944
CAS-Nummer: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR