AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid), Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
101500-1
101500-100
101500-50
101500-500
101500-1EA
1010
EUR
101500-1
101500.100
101500-50
101500.500
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid), Millipore®
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Formel:
C₈H₁₁ClFNO₂S Molecular Weight: 239.69 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00132962 CAS-Nummer: 30827-99-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...