Germanium 99,999 Metals basis, powder -100 mesh

Lieferant: Thermo Fisher Scientific

10190.09 10190.18
10190.09EA 233 EUR
10190.09 10190.18
Germanium 99,999 Metals basis, powder -100 mesh
Germanium

Purity: 99.999% CAS No.: 7440-56-4 Odour: Odourless MDL No.: MFCD00085310

Formel: Ge
Molecular Weight: 72.61 g/mol
Siedepunkt: 2830
Schmelzpunkt: 937.2
Lagertemperatur: Raumtemperatur
MDL: MFCD00085310
CAS-Nummer: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1 III
Merck Index: 12,04415

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR