Eisen 99,97 Metals basis, pieces, irregularly shaped ≤ 25 mm (1.0 in)
Lieferant: Thermo Fisher Scientific
10213.30
10213.A1
10213.A7
10213.PG_25KG
10213.30EA
48.6
EUR
10213.30
10213.A1
10213.A7
10213.PG_25KG
Eisen 99,97 Metals basis, pieces, irregularly shaped ≤ 25 mm (1.0 in)
Iron
Purity: 99.97+%
CAS No.: 7439-89-6
Physical Form: 90% 2.5-25mm (0.1-1.0in)
Appearance: Gray
MDL No.: MFCD00010999
Formel:
Fe Molecular Weight: 55.85 g/mol Siedepunkt: 3000 Schmelzpunkt: 1535 Dichte: 7.86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010999 CAS-Nummer: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...