di-Natriumhydrogenphosphat, wasserfrei ≥99,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals

102494CEA 68.1 EUR
102494C 102495D 10249LT
di-Natriumhydrogenphosphat, wasserfrei ≥99,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
di-Natriumhydrogenphosphat
Formel: Na₂HPO₄
Schmelzpunkt: 250 °C
Dichte: 1,53 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003496
CAS-Nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
pH (25°C; 5 %) 8.7 - 9.3
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 100 ppm
Loss on drying (105°C) Max. 0.2 %
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 50 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 100 ppm
Pb (Lead) Max. 10 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR