Wasser, AnalaR NORMAPUR® ISO 3696 Grade 3 zA

Lieferant: VWR Chemicals

Synonyme: Wasserstoffoxid, Hydrogenoxid

102923CEA 53.9 EUR
102923C 102927G 102928H 102922B 102926D
Wasser, AnalaR NORMAPUR® ISO 3696 Grade 3 zA
Wasser

Produziert durch Reversosmose und CDI

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Substances reducing KMnO4 Passes test
Leitfähigkeit (bei Abfüllung) Max. 1 µS/cm
TOC (bei Abfüllung) Max. 400 ppb
pH-Wert (25°C) 5,0 - 7,5
Verdampfungsrückstand Max. 1 ppm
Glührückstand Max. 1 ppm
Siliciumdioxid Max. 0,1 ppm
Cl (Chlorid) Max. 0.05 ppm
F (Fluorid) Max. 0,05 ppm
NH4 (Ammonium) Max. 0,2 ppm
NO3 (Nitrat) Max. 0.01 ppm
PO4 (Phosphat) Max. 0,01 ppm
SO4 (Sulfat) Max. 0,1 ppm
Ag (Silber) Max. 0,0004 ppm
Al (Aluminium) Max. 0.002 ppm
As (Arsen) Max. 0,002 ppm
Au (Gold) Max. 0,001 ppm
B (Bor) Max. 0.05 ppm
Ba (Barium) Max. 0.05 ppm
Be (Beryllium) Max. 0,002 ppm
Bi (Bismut) Max. 0,001 ppm
Ca (Calcium) Max. 0,05 ppm
Cd (Cadmium) Max. 0,001 ppm
Co (Cobalt) Max. 0,001 ppm
Cr (Chrom) Max. 0,0004 ppm
Cu (Kupfer) Max. 0,0004 ppm
Cu (Copper) Max. 0,001 ppm
Ga (Gallium) Max. 0,004 ppm
Ge (Germanium) Max. 0,001 ppm
In (Indium) Max. 0,001 ppm
K (Kalium) Max. 0,05 ppm
Li (Lithium) Max. 0,002 ppm
Mg (Magnesium) Max. 0,005 ppm
Mn (Mangan) Max. 0,0004 ppm
Mo (Molybdän) Max. 0,002 ppm
Na (Natrium) Max. 0,1 ppm
Ni (Nickel) Max. 0,0004 ppm
Pb (Blei) Max. 0,001 ppm
Pd (Palladium) Max. 0,008 ppm
Pt (Platin) Max. 0.001 ppm
Sb (Antimon) Max. 0.001 ppm
Sn (Zinn) Max. 0,001 ppm
Sr (Strontium) Max. 0,0004 ppm
Ti (Titan) Max. 0,001 ppm
Tl (Thallium) Max. 0,00005 ppm
V (Vanadium) Max. 0,001 ppm
Zn (Zink) Max. 0,004 ppm
Zr (Zirkon) Max. 0,001 ppm
ISO Standard Wasser Typ 3 Entspricht


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR