Praseodym 99% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.25 mm (0.01 in)
Lieferant: Thermo Fisher Scientific
10305.FF
10305.FFEA
466
EUR
10305.FF
Praseodym 99% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.25 mm (0.01 in)
Praseodymium
Purity: 99.5%
CAS No.: 7440-10-0
Physical Form: ≈1.27g/25x25mm, Packaged in mylar under argon
Appearance: Gray
Odour: Odourless
MDL No.: MFCD00011174
Formel:
Pr Molecular Weight: 140.91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011174 CAS-Nummer: 7440-10-0 EINECS: 231-120-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...