Diethyl-(trichlormethyl)phosphonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Diethyltrichloromethylphosphonate, (Trichloromethyl)phosphonic Acid Diethyl Ester
10306.04
10306.09
10306.04EA
49.9
EUR
10306.04
10306.09
Diethyl-(trichlormethyl)phosphonat ≥98%
Diethyl (trichloromethyl)phosphonate
Purity: 98%
CAS No.: 866-23-9
Molecular Formula: (C2H5O)2P(O)(CCl3)
Formula Weight: 255.48
Physical Form: Liquid
Density: 1.3670
Refractive Index: 1.4620
Boiling Point: 128°/14mm
UN No.: UN3278
MDL No.: MFCD00013666
Formel:
C₅H₁₀Cl₃O₃P Siedepunkt: 128 °C (14 mmHg) Dichte: 1,3670 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013666 CAS-Nummer: 866-23-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...