Amberlite® IR120 Wasserstoff-Form 620 - 830 μm (stark saurer Kationenaustauscher), Ionenaustauscher, Supelco®

Lieferant: Merck

10322
10322.EA 56.8 EUR
10322.
Amberlite® IR120 Wasserstoff-Form 620 - 830 μm (stark saurer Kationenaustauscher), Ionenaustauscher, Supelco®
Amberlite® IR120 Hydrogen form

Amberlite IR 120-H, gel type acidic cation exchange resin is of sulfonated polystyrene type having tremendous stability. It is an efficient heterogeneous catalyst for organic reactions.


Strongly acidic cation exchange resin suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc..

MDL: MFCD00132707
CAS-Nummer: 39389-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR