Natrium ≥99,95% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Bruchsiegelampulle
10336.03
10336.06
10336.03EA
153
EUR
10336.03
10336.06
Natrium ≥99,95% (Metall-Basis)
Sodium
Purity: 99.95%
CAS No.: 7440-23-5
Physical Form: Sealed in breakseal ampoules
Odour: Odourless
UN No.: 1428
MDL No.: MFCD00085307
Formel:
Na Molecular Weight: 22.99 g/mol Siedepunkt: 881 Schmelzpunkt: 97.8 Dichte: 0.9 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085307 CAS-Nummer: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: 4.3 I Merck Index: 13,08641 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...