Chlorbenzol ≥99,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Benzolchlorid, Phenylchlorid, Monochlorbenzol, Phenylchloride

103386DEA 173 EUR
103386D
Chlorbenzol ≥99,5%, AnalaR NORMAPUR® analytisches Reagens
Chlorbenzol
Formel: C₆H₅Cl
Molecular Weight: 112,56 g/mol
Siedepunkt: 130 °C (1013 hPa)
Schmelzpunkt: –45 °C
Dichte: 1,11 g/cm³ (20 °C)
Flammpunkt: 28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000530
CAS-Nummer: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.5 %
IR Spectrum Passes test
Colouration Max. 10 APHA
Density (20/4) 1.100 - 1.110
Evaporation residue Max. 0.05 %
Water Max. 0.05 %
Al (Aluminium) Max. 2.0 ppm
Ba (Barium) Max. 1.0 ppm
Ca (Calcium) Max. 5.0 ppm
Cd (Cadmium) Max. 0.5 ppm
Co (Cobalt) Max. 0.2 ppm
Cr (Chromium) Max. 0.2 ppm
Cu (Copper) Max. 0.2 ppm
Fe (Iron) Max. 2.0 ppm
K (Potassium) Max. 1.0 ppm
Mg (Magnesium) Max. 0.5 ppm
Mn (Manganese) Max. 0.2 ppm
Na (Sodium) Max. 5.0 ppm
Ni (Nickel) Max. 0.2 ppm
Pb (Lead) Max. 0.5 ppm
Sr (Strontium) Max. 0.2 ppm
Zn (Zinc) Max. 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR