Amberlite® XAD®-2, Porengröße: 90 Å mean 20 - 60 mesh Ionenaustauscherharz, Supelco®
Lieferant: Merck
10357
10357.EA
142
EUR
10357.
Amberlite® XAD®-2, Porengröße: 90 Å mean 20 - 60 mesh Ionenaustauscherharz, Supelco®
Amberlite® XAD®-2
Amberlite® XAD-2, a hydrophobic copolymer of styrene-divinylbenzene resin, requires longer purification process and is commonly used as a sorbent. Amberlite® XAD-2 resin sorbs organic compounds depending upon the aromaticity and size of the hydrophobic groups through low-energy physical interactions.
- Surface area: ~300 m²/g
- Used as adsorbent in the extraction of organochlorine compounds using Soxhlet extraction technique
- Used as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
CAS-Nummer:
9003-70-7 |