D(-)-Fructose 99,0, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals

Synonyme: Lävulose, Schleimzucker, D(-)-Lävulose, Fruchtzucker

103674YEA 56.7 EUR
103674Y
D(-)-Fructose 99,0, AnalaR NORMAPUR® zA
D(-)-Fructose
Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Siedepunkt: 552
Schmelzpunkt: 100…110 °C
Dichte: 1.59 g/cm³ (20 °C. OECD 109)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148910
CAS-Nummer: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated on anhydrous) Min. 99.0 %
IR Spectrum Passes test
Spec.opt.rot.(10 %;water)(on anhydrous) -93.5 to -91.0 °
Glucose Max. 0.5 %
Ignition residue (SO4) Max. 0.05 %
Insolubility in water Max. 50 ppm
Water Max. 0.5 %
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 1 ppm
Cd (Cadmium) Max. 1 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 1 ppm
Zn (Zinc) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR