Ytterbium 99,9 REO basis, foil, Dicke 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific

10426.FF 10426.FI
10426.FFEA 211 EUR
10426.FF 10426.FI
Ytterbium 99,9 REO basis, foil, Dicke 1.0 mm (0.04 in)
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Physical Form: ≈4.4g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011286

Formel: Yb
Molecular Weight: 173.04 g/mol
Siedepunkt: 1427
Schmelzpunkt: 824
Dichte: 6.977 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011286
CAS-Nummer: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR